Thiophene-3-thiocarboxamide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Thiophene-3-carbothioic acid amide, 3-Carbamothioylthiophene
OR25930-1G
OR25930-5G
APOSOR25930-1GEA
112
GBP
APOSOR25930-1G
APOSOR25930-5G
Thiophene-3-thiocarboxamide
Thiophene-3-thiocarboxamide
Formula:
C₅H₅NS₂ MW: 143,23 g/mol Melting Pt: 109…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173746 CAS Number: 24044-76-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...