5-(Phenylethynyl)nicotinic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-(Phenylethynyl)pyridine-3-carboxylic acid
OR25628-1G
OR25628-5G
OR25628-100MG
APOSOR25628-1GEA
119
GBP
APOSOR25628-1G
APOSOR25628-5G
APOSOR25628-100MG
5-(Phenylethynyl)nicotinic acid
5-(Phenylethynyl)nicotinic acid
Formula:
C₁₄H₉NO₂ MW: 223,227 g/mol Melting Pt: 245…247 °C Storage Temperature: Ambient |
MDL Number:
MFCD00174055 CAS Number: 175203-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...