3-Ethoxy-2-[(naphth-2-yl)sulphonyl]acrylonitrile

Supplier: Apollo Scientific

Synonyms: 3-Ethoxy-2-[(naphth-2-yl)sulphonyl]prop-2-enenitrile

OR25090-1G OR25090-5G
APOSOR25090-1GEA 78.5 GBP
APOSOR25090-1G APOSOR25090-5G
3-Ethoxy-2-[(naphth-2-yl)sulphonyl]acrylonitrile
3-Ethoxy-2-[(naphth-2-yl)sulphonyl]acrylonitrile
Formula: C₁₅H₁₃NO₃S
MW: 287.334 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067880
CAS Number: 32083-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR