3-Methyl-5-phenylisoxazole-4-carbonyl chloride

Supplier: Apollo Scientific
Danger

Synonyms: 4-(Chlorocarbonyl)-3-methyl-5-phenylisoxazole

OR23336-1G OR23336-250MG
APOSOR23336-1GEA 43.4 GBP
APOSOR23336-1G APOSOR23336-250MG
3-Methyl-5-phenylisoxazole-4-carbonyl chloride
3-Methyl-5-phenylisoxazole-4-carbonyl chloride
Formula: C₁₁H₈ClNO₂
MW: 221,64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03086121
CAS Number: 91182-77-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR