3,4-Dihydro-2H-1,5-benzodioxepin-7-boronic acid 90%

Supplier: Apollo Scientific
Danger

OR23213-1G OR23213-250MG
APOSOR23213-1GEA 238 GBP
APOSOR23213-1G APOSOR23213-250MG
3,4-Dihydro-2H-1,5-benzodioxepin-7-boronic acid 90%
3,4-Dihydro-2H-1,5-benzodioxepin-7-boronic acid
Formula: C₉H₁₁BO₄
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD02681910
CAS Number: 279261-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR