4-Bromo-3-methylphenacyl bromide, tech
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-1-(4-bromo-3-methylphenyl)ethanone
OR18870-1G
OR18870-5G
APOSOR18870-1GEA
206
GBP
APOSOR18870-1G
APOSOR18870-5G
4-Bromo-3-methylphenacyl bromide, tech
4-Bromo-3-methylphenacyl bromide
Formula:
C₉H₈Br₂O MW: 291,967 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20266827 CAS Number: 3114-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...