Sodium 4-chlorobenzoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chlorobenzoic acid sodium salt

OR1870X-25G OR1870X-5G OR1870X-100G
APOSOR1870X-25GEA 0 GBP
APOSOR1870X-25G APOSOR1870X-5G APOSOR1870X-100G
Sodium 4-chlorobenzoate 98%
Sodium 4-chlorobenzoate
Formula: C₇H₄ClO₂·Na
MW: 178,548 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016523
CAS Number: 3686-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR