Methyl 3-amino-4-iodobenzothiophene-2-carboxylate

Supplier: Apollo Scientific

OR183427-1G OR183427-250MG
APOSOR183427-1GEA 328 GBP
APOSOR183427-1G APOSOR183427-250MG
Methyl 3-amino-4-iodobenzothiophene-2-carboxylate
Methyl 3-amino-4-iodobenzothiophene-2-carboxylate
Formula: C₁₀H₈INO₂S
MW: 333,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29163652
CAS Number: 1603418-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR