2-Bromo-5-nitrophenyl methyl sulphone 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-2-(methylsulphonyl)-4-nitrobenzene
OR17674-1G
OR17674-5G
OR17674-250MG
OR17674-25G
APOSOR17674-1GEA
315
GBP
APOSOR17674-1G
APOSOR17674-5G
APOSOR17674-250MG
APOSOR17674-25G
2-Bromo-5-nitrophenyl methyl sulphone 95%
2-Bromo-5-nitrophenyl methyl sulphone
Formula:
C₇H₆BrNO₄S MW: 280,096 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11052985 CAS Number: 1048916-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...