3-Methyl-2-nitrobenzaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Nitro-m-tolualdehyde, 3-Formyl-2-nitrotoluene
OR16689-1G
OR16689-250MG
OR16689-5G
APOSOR16689-1GEA
178
GBP
APOSOR16689-1G
APOSOR16689-250MG
APOSOR16689-5G
3-Methyl-2-nitrobenzaldehyde 97%
3-Methyl-2-nitrobenzaldehyde
Formula:
C₈H₇NO₃ Melting Pt: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD08703364 CAS Number: 5858-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...