(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid

Supplier: Apollo Scientific
Warning

Synonyms: (R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen, (2R)-2-(6-Methoxynaphth-2-yl)propionic acid, R-Naproxen, 2-[(1R)-1-Carboxyethyl]-6-methoxynaphthalene

OR16409-1G OR16409-250MG OR16409-5G
APOSOR16409-1GEA 98.3 GBP
APOSOR16409-1G APOSOR16409-250MG APOSOR16409-5G
(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid
(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid
Formula: C₁₄H₁₄O₃
MW: 230,259 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00870716
CAS Number: 23979-41-1
EINECS: 245-966-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR