Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3-Bis(methoxycarbonyl)pyrazolo[1,5-a]pyridine
OR14497-1G
OR14497-5G
APOSOR14497-1GEA
65.6
GBP
APOSOR14497-1G
APOSOR14497-5G
Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate
Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate
Formula:
C₁₁H₁₀N₂O₄ MW: 234,208 g/mol Boiling Pt: 174…178 °C (0,23 mm Hg) Melting Pt: 66…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD06411548 CAS Number: 5825-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...