4'-Hydroxy-4-methoxychalcone

Supplier: Apollo Scientific

OR14434-25G OR14434-50G
APOSOR14434-25GEA 516 GBP
APOSOR14434-25G APOSOR14434-50G
4'-Hydroxy-4-methoxychalcone
4'-Hydroxy-4-methoxychalcone
Formula: C₁₆H₁₄O₃
MW: 254,281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020170
CAS Number: 6338-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR