2-(Bromomethyl)-6-(pyrrolidin-1-yl)pyridine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-[6-(Bromomethyl)pyridin-2-yl]pyrrolidine
OR12395-1G
OR12395-250MG
APOSOR12395-1GEA
414
GBP
APOSOR12395-1G
APOSOR12395-250MG
2-(Bromomethyl)-6-(pyrrolidin-1-yl)pyridine 97%
2-(Bromomethyl)-6-(pyrrolidin-1-yl)pyridine
Formula:
C₁₀H₁₃BrN₂ MW: 241,128 g/mol Melting Pt: 69…72,5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09817465 CAS Number: 869901-04-2 UN: 2923 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...