(E)-3-(Naphth-1-yl)acrylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2E)-3-(Naphth-1-yl)prop-2-enoic acid, (E)-3-(Naphthalen-1-yl)acrylic acid, trans-3-(Naphth-1-yl)acrylic acid
OR12209-10G
OR12209-50G
APOSOR12209-10GEA
132
GBP
APOSOR12209-10G
APOSOR12209-50G
(E)-3-(Naphth-1-yl)acrylic acid
(E)-3-(Naphth-1-yl)acrylic acid
Formula:
C₁₃H₁₀O₂ MW: 198,217 g/mol Melting Pt: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014317 CAS Number: 2006-14-6 EINECS: 235-887-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...