Methyl 3-amino-7-cyano-1-benzothiophene-2-carboxylate

Supplier: Apollo Scientific
Warning

OR111211-1G OR111211-5G
APOSOR111211-1GEA 17.1 GBP
APOSOR111211-1G APOSOR111211-5G
Methyl 3-amino-7-cyano-1-benzothiophene-2-carboxylate
Methyl 3-amino-7-cyano-1-benzothiophene-2-carboxylate
Formula: C₁₁H₈N₂O₂S
MW: 232,258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04110076
CAS Number: 181283-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR