(z)-Ethyl 2-chloro-2-(2-(4-chlorophenyl)hydrazono)acetate
Supplier: Apollo Scientific
OR111076-10G
OR111076-1G
OR111076-5G
APOSOR111076-10GEA
735
GBP
APOSOR111076-10G
APOSOR111076-1G
APOSOR111076-5G
(z)-Ethyl 2-chloro-2-(2-(4-chlorophenyl)hydrazono)acetate
(z)-Ethyl 2-chloro-2-(2-(4-chlorophenyl)hydrazono)acetate
Formula:
C₁₀H₁₀Cl₂N₂O₂ MW: 261,105 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00228728 CAS Number: 27143-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...