2-Tetrahydro-2H-pyran-4-yl-1,3,4-oxadiazole

Supplier: Apollo Scientific
Warning

OR111067-1G OR111067-5G
APOSOR111067-1GEA 254 GBP
APOSOR111067-1G APOSOR111067-5G
2-Tetrahydro-2H-pyran-4-yl-1,3,4-oxadiazole
2-Tetrahydro-2H-pyran-4-yl-1,3,4-oxadiazole
Formula: C₇H₁₀N₂O₂
MW: 154,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27997558
CAS Number: 1082413-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR