5-(1-Chloro-2-methylpropan-2-yl)-3-(thiophen-2-yl)-1,2,4-oxadiazole
Supplier: Apollo Scientific
OR110778-1G
OR110778-5G
APOSOR110778-1GEA
253
GBP
APOSOR110778-1G
APOSOR110778-5G
5-(1-Chloro-2-methylpropan-2-yl)-3-(thiophen-2-yl)-1,2,4-oxadiazole
5-(1-Chloro-2-methylpropan-2-yl)-3-(thiophen-2-yl)-1,2,4-oxadiazole
Formula:
C₁₀H₁₁ClN₂OS MW: 242,725 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23159170 CAS Number: 1427460-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...