Methyl 4-octylbenzoate

Supplier: Apollo Scientific

OR110469-10G OR110469-1G OR110469-5G
APOSOR110469-10GEA 391 GBP
APOSOR110469-10G APOSOR110469-1G APOSOR110469-5G
Methyl 4-octylbenzoate
Methyl 4-octylbenzoate
Formula: C₁₆H₂₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06203813
CAS Number: 54256-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR