Ethyl 2-methyl-3-nitrobenzoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(Ethoxycarbonyl)-2-methylnitrobenzene, Ethyl 3-nitro-o-toluate, 2-(Ethoxycarbonyl)-6-nitrotoluene
OR110420-25G
OR110420-100G
APOSOR110420-25GEA
13.2
GBP
APOSOR110420-25G
APOSOR110420-100G
Ethyl 2-methyl-3-nitrobenzoate
Ethyl 2-methyl-3-nitrobenzoate
Formula:
C₁₀H₁₁NO₄ MW: 209,199 g/mol Melting Pt: 34…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD07359723 CAS Number: 59382-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...