3-Bromo-1H-indazol-7-amine

Supplier: Apollo Scientific
Warning

OR110360-1G OR110360-250MG
APOSOR110360-1GEA 205 GBP
APOSOR110360-1G APOSOR110360-250MG
3-Bromo-1H-indazol-7-amine
3-Bromo-1H-indazol-7-amine
Formula: C₇H₆BrN₃
MW: 212,047 g/mol
Melting Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD09027010
CAS Number: 316810-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR