Ethyl 1-methyl-1H-pyrrole-2-carboxylate

Supplier: Apollo Scientific

OR110264-1G OR110264-5G
APOSOR110264-1GEA 84 GBP
APOSOR110264-1G APOSOR110264-5G
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Formula: C₈H₁₁NO₂
MW: 153,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179573
CAS Number: 23466-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR