4-Nitropyridine 95%
Supplier: Apollo Scientific
OR10961-1G
OR10961-250MG
OR10961-5G
APOSOR10961-1GEA
87.8
GBP
APOSOR10961-1G
APOSOR10961-250MG
APOSOR10961-5G
4-Nitropyridine 95%
4-Nitropyridine
Formula:
C₅H₄N₂O₂ MW: 124,097 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00054554 CAS Number: 1122-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...