{3-[(Methylsulphonyl)amino]phenyl}acetic acid

Supplier: Apollo Scientific
Warning

Synonyms: N-[(3-Carboxymethyl)phenyl]methanesulphonamide

OR10893-500MG OR10893-50MG
APOSOR10893-500MGEA 218 GBP
APOSOR10893-500MG APOSOR10893-50MG
{3-[(Methylsulphonyl)amino]phenyl}acetic acid
{3-[(Methylsulphonyl)amino]phenyl}acetic acid
Formula: C₉H₁₁NO₄S
MW: 229,253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411194
CAS Number: 407640-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR