1-Hexanoylpiperazine

Supplier: Apollo Scientific
Warning

Synonyms: 1-(1-Piperazinyl)-1-hexanone

OR0839-1G OR0839-5G
APOSOR0839-1GEA 119 GBP
APOSOR0839-1G APOSOR0839-5G
1-Hexanoylpiperazine
1-Hexanoylpiperazine
Formula: C₁₀H₂₀N₂O
MW: 184,279 g/mol
Boiling Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD04973345
CAS Number: 18903-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR