6-Chloro-2-phenylimidazo[1,2-b]pyridazine
Supplier: Apollo Scientific
|
Warning
|
OR01789-1G
OR01789-250MG
APOSOR01789-1GEA
414
GBP
APOSOR01789-1G
APOSOR01789-250MG
6-Chloro-2-phenylimidazo[1,2-b]pyridazine
6-Chloro-2-phenylimidazo[1,2-b]pyridazine
Formula:
C₁₂H₈ClN₃ MW: 229,665 g/mol Melting Pt: 195…200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03407868 CAS Number: 1844-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...