N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide

Supplier: Apollo Scientific

OR015042-1G OR015042-250MG
APOSOR015042-1GEA 46 GBP
APOSOR015042-1G APOSOR015042-250MG
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide
Formula: C₁₄H₂₂N₄O
MW: 262,351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12457658
CAS Number: 262368-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR