Sarsasapogenin
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(25S)-Spirostan-3β-ol, Parigenin, (2aR,2'R,4S,5'S,6aS,6bS,8aS,8bR,9S,11aS,12aS,12bR)-5',6a,8a,9-Tetramethyldocosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol
BITH3027-100MG
BITH3027-25MG
APOSBITH3027-100MGEA
89.2
GBP
APOSBITH3027-100MG
APOSBITH3027-25MG
Sarsasapogenin
Sarsasapogenin
Formula:
C₂₇H₄₄O₃ MW: 416,636 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00270414 CAS Number: 126-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...