2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide

Supplier: Apollo Scientific
Warning

Synonyms: 2,3,4,6-Tetra-O-pivaloyl-alpha-D-glucopyranosyl bromide, (2R,3R,4S,5R,6R)-2-Bromo-6-((pivaloyloxy)methyl)tetrahydro-2H-pyran-3,4,5-triyl tris(2,2-dimethylpropanoate)

BIT1007-25G BIT1007-100G BIT1007-50G
APOSBIT1007-25GEA 45.9 GBP
APOSBIT1007-25G APOSBIT1007-100G APOSBIT1007-50G
2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide
2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide
Formula: C₂₆H₄₃BrO₉
MW: 579.518 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08275217
CAS Number: 81058-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR