Dicamba-[13C6]
Supplier: Apollo Scientific
|
Danger
|
BISC1013-10MG
BISC1013-5MG
BISC1013-2MG
APOSBISC1013-10MGEA
2700
GBP
APOSBISC1013-10MG
APOSBISC1013-5MG
APOSBISC1013-2MG
Dicamba-[13C6]
Dicamba-[13C6]
Formula:
[₁₃C]₆C₂H₆Cl₂O₃ MW: 226,993 g/mol Melting Pt: 112…116 °C Storage Temperature: Freezer |
MDL Number:
MFCD11616810 CAS Number: 1173023-06-7 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...