4-Methylumbelliferyl α-L-arabinofuranoside
Supplier: Apollo Scientific
|
Warning
|
BIM2041-25MG
BIM2041-50MG
APOSBIM2041-25MGEA
112
GBP
APOSBIM2041-25MG
APOSBIM2041-50MG
4-Methylumbelliferyl α-L-arabinofuranoside
4-Methylumbelliferyl α-L-arabinofuranoside
Formula:
C₁₅H₁₆O₇ MW: 308,283 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00057308 CAS Number: 77471-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...