Disodium 4,4'-diisothiocyanato-2,2'-stilbenedisulphonate hydrate

Supplier: Apollo Scientific
Danger

Synonyms: 4,4'-Diisothiocyanatostilbene-2,2'-disulphonic acid disodium salt hydrate

BID2117-100MG BID2117-1G BID2117-500MG
APOSBID2117-100MGEA 36.8 GBP
APOSBID2117-100MG APOSBID2117-1G APOSBID2117-500MG
Disodium 4,4'-diisothiocyanato-2,2'-stilbenedisulphonate hydrate
Disodium 4,4'-diisothiocyanato-2,2'-stilbenedisulphonate hydrate

A negatively charged homobifunctional cross-linkingreagent. Can be used in cell transport studies.

Formula: C₁₆H₈N₂O₆S₄·2Na·H₂O
MW: 516,499 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00149576
CAS Number: 207233-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR