4-Cyclohexylbutyl β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
BICL4302-1G
BICL4302-500MG
APOSBICL4302-1GEA
1010
GBP
APOSBICL4302-1G
APOSBICL4302-500MG
4-Cyclohexylbutyl β-D-glucopyranoside ≥98%
4-Cyclohexylbutyl β-D-glucopyranoside
Storage Temperature:
Ambient |
CAS Number:
869542-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...