Epimaltose ≥95%

Supplier: Apollo Scientific

Synonyms: 4-O-β-D-Glucopyranosyl-D-mannopyranose

BICL4296-1MG BICL4296-2MG
APOSBICL4296-1MGEA 656 GBP
APOSBICL4296-1MG APOSBICL4296-2MG
Epimaltose ≥95%
Epimaltose
Formula: C₁₂H₂₂O₁₁
MW: 342,296 g/mol
Storage Temperature: Ambient
CAS Number: 70832-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR