2,5-Anhydro-D-mannitol ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: NSC 129241

BICL4243-100MG BICL4243-250MG BICL4243-500MG
APOSBICL4243-100MGEA 328 GBP
APOSBICL4243-100MG APOSBICL4243-250MG APOSBICL4243-500MG
2,5-Anhydro-D-mannitol ≥98%
2,5-Anhydro-D-mannitol
Formula: C₆H₁₂O₅
MW: 164,156 g/mol
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00012443
CAS Number: 41107-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR