2-O-(α-L-Fucopyranosyl)-D-galactose ≥98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-O-(6-deoxy-α-L-galactopyranosyl)-D-galactose, Blood Group H disaccharide
BICL4235-10MG
BICL4235-5MG
BICL4235-1MG
APOSBICL4235-10MGEA
858
GBP
APOSBICL4235-10MG
APOSBICL4235-5MG
APOSBICL4235-1MG
2-O-(α-L-Fucopyranosyl)-D-galactose ≥98%
2-O-(α-L-Fucopyranosyl)-D-galactose
Formula:
C₁₂H₂₂O₁₀ MW: 326,297 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD07357245 CAS Number: 24656-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...