Methyl 1,2,3,4-tetra-O-pivaloyl-β-D-glucopyranosyluronate ≥98%

Supplier: Apollo Scientific

Synonyms: 1,2,3,4-Tetra-O-pivaloyl-β-D-glucuronic acid methyl ester

BICL4142-1G BICL4142-250MG BICL4142-500MG
APOSBICL4142-1GEA 1150 GBP
APOSBICL4142-1G APOSBICL4142-250MG APOSBICL4142-500MG
Methyl 1,2,3,4-tetra-O-pivaloyl-β-D-glucopyranosyluronate ≥98%
Methyl 1,2,3,4-tetra-O-pivaloyl-β-D-glucopyranosyluronate
Formula: C₂₇H₄₄O₁₁
MW: 544,632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145134
CAS Number: 86448-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR