Methyl 2,3,4-tri-O-acetyl-1-O-trichloroacetimidoyl-α-D-glucopyranosyluronate ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: (2S,3S,4S,5R,6R)-2-(Methoxycarbonyl)-6-(2,2,2-trichloro-1-iminoethoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate, 2,3,4-Tri-O-acetyl-α-D-glucuronic acid methyl ester trichloroacetimidate

BICL4140-100MG BICL4140-500MG BICL4140-250MG
APOSBICL4140-100MGEA 202 GBP
APOSBICL4140-100MG APOSBICL4140-500MG APOSBICL4140-250MG
Methyl 2,3,4-tri-O-acetyl-1-O-trichloroacetimidoyl-α-D-glucopyranosyluronate ≥99%
Methyl 2,3,4-tri-O-acetyl-1-O-trichloroacetimidoyl-α-D-glucopyranosyluronate
Formula: C₁₅H₁₈Cl₃NO₁₀
MW: 478,6600 g/mol
Melting Pt: 125…126 °C
Storage Temperature: Freezer
MDL Number: MFCD02094293
CAS Number: 92420-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR