1,2-O-Isopropylidene-β-L-idofuranuronic acid, γ-lactone ≥99%

Supplier: Apollo Scientific

Synonyms: β-L-Idofurano-6,3-lactone acetonide, β-L-Iduronolactone acetonide

BICL4122-1G BICL4122-250MG BICL4122-500MG
APOSBICL4122-1GEA 632 GBP
APOSBICL4122-1G APOSBICL4122-250MG APOSBICL4122-500MG
1,2-O-Isopropylidene-β-L-idofuranuronic acid, γ-lactone ≥99%
1,2-O-Isopropylidene-β-L-idofuranuronic acid, γ-lactone
Storage Temperature: Ambient
CAS Number: 29514-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR