Benzyl 2,3-O-isopropylidene-6-O-tosyl-α-L-sorbofuranoside ≥99%
Supplier: Apollo Scientific
BICL4051-500MG
BICL4051-100MG
BICL4051-250MG
APOSBICL4051-500MGEA
909
GBP
APOSBICL4051-500MG
APOSBICL4051-100MG
APOSBICL4051-250MG
Benzyl 2,3-O-isopropylidene-6-O-tosyl-α-L-sorbofuranoside ≥99%
Benzyl 2,3-O-isopropylidene-6-O-tosyl-α-L-sorbofuranoside
Storage Temperature:
Ambient |
CAS Number:
1459288-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...