2,6-Anhydro-3-deoxy-D-glycero-D-galacto-non-2-enoic acid ≥98%
Supplier: Apollo Scientific
Synonyms:
Kdn-2-en
BICL4029-1MG
BICL4029-5MG
BICL4029-2MG
APOSBICL4029-1MGEA
223
GBP
APOSBICL4029-1MG
APOSBICL4029-5MG
APOSBICL4029-2MG
2,6-Anhydro-3-deoxy-D-glycero-D-galacto-non-2-enoic acid ≥98%
2,6-Anhydro-3-deoxy-D-glycero-D-galacto-non-2-enoic acid
Formula:
C₉H₁₄O₈ MW: 250,203 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683259 CAS Number: 188854-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...