3-Indolyl butyrate

Supplier: Apollo Scientific

BIB6080-5G BIB6080-10G BIB6080-25G
APOSBIB6080-5GEA 380 GBP
APOSBIB6080-5G APOSBIB6080-10G APOSBIB6080-25G
3-Indolyl butyrate
3-Indolyl butyrate
Formula: C₁₂H₁₃NO₂
MW: 203.237 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00022718
CAS Number: 4346-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR