Haematoxylin Gill No. 2, Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
|
Danger
|
Synonyms:
Hydroxybrasilin
, DcH-48
, Natural Black I
GHS232-1LEA
80.5
GBP
GHS232-1L
Haematoxylin Gill No. 2, Sigma-Aldrich®
Haematoxylin staining solutions
Gill formulation No. 2 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.
Formula:
C₁₆H₁₄O₆ MW: 302,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...