Trilostane

Supplier: Thermo Fisher Scientific
Warning

464380010 464382500
ACRO464380010EA 516 GBP
ACRO464380010 ACRO464382500
Trilostane
Trilostane

  • Bioactive

Formula: C₂₀H₂₇NO₃
Melting Pt: 284 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00199295
CAS Number: 13647-35-3

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR