VWR® Advanced Hotplates
Supplier: VWR Collection
444-0589EA 613 GBP
444-0589 444-0590 444-0592 444-0586 444-0587 444-0575 444-0577 444-0578 444-0580 444-0581 444-0583 444-0584
VWR® Advanced Hotplates
Heaters Hotplates
VWR® Advanced hotplates deliver accurate and repeatable results. Units are microprocessor controlled and have an easy to read LED display for set point temperature. The control panel features easy to use controls which allow users to dial in temperature adjustments. Rear housing features an off-centred, integral support rod holder with locking knob to accept the optional support rod and clamp kit. The low profile design takes up less space and fits into fume hoods. A spill resistant housing channels fluids away from internal components.
  • Excellent temperature uniformity
  • Microprocessor controlled with built-in memory allowing recall of last set temperature (even if unit has been turned off)
  • Cool touch, chemically resistant housing
  • LED display for temperature
  • Choice of models with ceramic or aluminium top plates
A 'hot' symbol warning light is illuminated when heat is turned on and remains on until top plate cools down. Enhanced electronics regulate heating and bring samples to temperature quickly and efficiently. Temperature measurement is controlled more accurately by using a resistance thermometer (RTD). Ceramic tops feature a chemically resistant, reflective white top plate surface that is easy to clean. Durable aluminium tops will not crack or chip, and provide a more even heating surface.
Ordering information: Optional support rod kit is available which includes a 457 mm stainless steel support rod, thermometer/temperature probe extension clamp, three-prong medium swivel clamp, and hook connector.
Caution: * RT= Ambient.