Saccharin ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Synonyms:
o-Sulphobenzimide, o-Benzoic sulphimide
87073.260EA
63.4
GBP
87073.260
87073.360
Saccharin ≥99%, GPR RECTAPUR®
Saccharin
Formula:
C₇H₅NO₃S |
MDL Number:
MFCD00005866 CAS Number: 81-07-2 EINECS: 201-321-0 |
Specification Test Results
Assay | Min. 99 % |
Melting point | 226 --> 230 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...