Diisobutylaluminium hydride 1M in hexane

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

Packaged under Nitrogen in resealable AcroSeal® bottles

H37222.9Z H37222.AE
H37222.9ZEA 0 GBP
H37222.9Z H37222.AE
Diisobutylaluminium hydride 1M in hexane
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Melting Pt: -80
Density: 0.798
Flash Pt: –23°C (–9°F)
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.2 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR