2-Acetylnaphthalene 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone

158881000 158885000
ACRO158881000EA 59.3 GBP
ACRO158881000 ACRO158885000
2-Acetylnaphthalene 99%
2-Acetylnaphthalene
Formula: C₁₂H₁₀O
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 53…56 °C
Density: 1,1171 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥98.5 %
Color scale ≤20 APHA
Ash ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR