Triphenylphosphine sulphide ≥98%
Supplier: Thermo Scientific
|
Danger
|
A16884.14
A16884.22
A16884.14EA
53.3
GBP
A16884.14
A16884.22
Triphenylphosphine sulphide ≥98%
Triphenylphosphine sulphide
Purity: 98%
CAS No.: 3878-45-3
Molecular Formula: C18H15PS
Formula Weight: 294.36
Melting Point: 158-164°
Beilstein Reference: 645872
UN No.: 3464
MDL No.: MFCD00004917
Formula:
C₁₈H₁₅PS MW: 294,36 g/mol Melting Pt: 158…164 °C Density: 1,191 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004917 CAS Number: 3878-45-3 EINECS: 223-407-7 UN: 3464 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...